Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526699
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526699 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526699 Supplier Invitrogen™ Supplier No. PA526699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytokeratin 1 Polyclonal Antibody for Western Blot, IHC (P), Flow

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between approximately40 kDa and approximately68 kDa and isoelectric pH between 4. 9 - 7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene KRT1
Gene Accession No. P04264
Gene Alias 67 kDa cytokeratin; CK1; CK-1; cytokeratin 1; cytokeratin-1; EHK; EHK1; epidermolytic hyperkeratosis 1; epithelial keratin 1; Epithelial keratin-1; EPPK; Hair alpha protein; K1; Kb1; KER1; keratin 1; keratin 1 (epidermolytic hyperkeratosis); keratin 1, type II; keratin 86; keratin complex 2, basic, gene 1; keratin, type II cytoskeletal 1; Keratin-1; KRT1; krt1 {ECO:0000250; KRT1A; Krt-2.1; Krt2-1; Krt86; KRTA; NEPPK; type II keratin Kb1; type-II keratin Kb1; UniProtKB:P04264}
Gene Symbols KRT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 415-443 amino acids from the central region of human KRT1.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3848
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.