Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 10/13 Antibody (SPM262), PE , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311558PE
Description
Cytokeratin 10/13 Monoclonal antibody specifically detects Cytokeratin 10/13 in Human, Feline samples. It is validated for Flow CytometrySpecifications
Cytokeratin 10/13 | |
Monoclonal | |
PE | |
BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, keratin, type I cytoskeletal 10, keratin-10, KPP | |
Cytoskeletal preparation extracted from human ectocervical epithelium | |
0.1 mL | |
Cell Biology, Cellular Markers | |
3858 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry | |
SPM262 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Feline | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction