Learn More
Invitrogen™ Cytokeratin 10/13 Monoclonal Antibody (EBS-IF-003)

Description
EBS-IF-003 reacts with 53kDa (CK13) and 56.6kDa (CK10) cytokeratin proteins as indicated by immunoblotting. On formalin-fixed, paraffin-embedded tissue sections EBS-IF-003 recognizes only CK13. On cryostat sections EBS-IF-003 serves as differentiation-related marker of all stratified epithelia; it stains all suprabasal cells in both cornifying and noncornifying stratified epithelia and more differentiated cells of squamous carcinomas. In paraffin sections EBS-IF-003 does not stain CK10 positive, CK13 negative epithelia, as for example epidermis.
Specifications
Specifications
| Antigen | Cytokeratin 10/13 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | EBS-IF-003 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | KRT10 |
| Gene Accession No. | P13645, P13646, Q6EIZ0 |
| Gene Alias | BCIE; BIE; CK10; CK-10; CK13; CK-13; cytokeratin 10; cytokeratin 13; cytokeratin-10; Cytokeratin-13; EHK; epithelial keratin 10; K10; K13; Ker10; keratin 10; keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin 13; keratin 13, type I; keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 13; keratin-10; Keratin-13; KPP; KRT10; KRT13; WSN2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.