Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 10 Antibody (1C3D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | Cytokeratin 10 |
---|---|
Clone | 1C3D9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:200-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61737
![]() |
Novus Biologicals
NBP261737 |
0.1 mL |
Each of 1 for $560.50
|
|
|||||
Description
Cytokeratin 10 Monoclonal specifically detects Cytokeratin 10 in Human, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 10 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Monkey | |
3858 | |
Purified recombinant fragment of human KRT10 (AA: 345-454 ) expressed in E. Coli. | |
Primary | |
176 kDa |
1C3D9 | |
Western Blot 1:200-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Monoclonal | |
Purified | |
Cytoskeleton Markers | |
BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
KRT10 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title