Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 10 Monoclonal Antibody (ZY420-LH1), Abbomax

Catalog No. 502204451
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4451 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4451 Supplier Abbomax Inc Supplier No. 606490
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Breast carcinoma Cellular location: Membrane and cytoplsm.

Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Cytokeratin 10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ZY420-LH1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene KRT10
Gene Accession No. P02535, P13645, Q6IFW6
Gene Alias 56 kDa cytokeratin; BCIE; BIE; CK10; CK-10; CK-17; cytokeratin 10; cytokeratin-10; cytokeratin-17; D130054E02Rik; EHK; epidermal keratin 10; epithelial keratin 10; intermediate filament protein; K10; K1C1; Ka10; Ker10; keratin 10; keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin complex 1, acidic, gene 10; keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 59 kDa; keratin-10; keratin-17; KPP; Krt-1.10; KRT10; krt10 {ECO:0000250; Krt1-10; suprabasal cytokeratin 10; type I keratin KA10; UniProtKB:P13645}
Host Species Mouse
Immunogen A synthetic peptide corresponding to the C-terminus of CK19
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16661, 3858, 450225
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.