Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 10 Antibody (RM386), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289092
Description
Cytokeratin 10 Monoclonal specifically detects Cytokeratin 10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 10 | |
Monoclonal | |
Unconjugated | |
PBS, 50% Glycerol, 1% BSA | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM386 | |
Western Blot 1:200 - 1:1000, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin | |
BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
A peptide corresponding to the C-terminus of human Cytokeratin 10 | |
100 μL | |
Cell Biology, Cellular Markers | |
3858 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction