Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 10 Rabbit anti-Human, Mouse, Rat, Clone: 4S5Y7, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
| Antigen | Cytokeratin 10 |
|---|---|
| Clone | 4S5Y7 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
Description
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| Cytokeratin 10 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
| A synthetic peptide corresponding to a sequence within amino acids 485-584 of human Cytokeratin 10 (P13645). GGHGGGHGGSSGGGYGGGSSGGGSSGGGYGGGSSSGGHGGSSSGGYGGGSSGGGGGGYGGGSSGGGSSSGGGYGGGSSSGGHKSSSSGSVGESSSKGPRY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 4S5Y7 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 3858 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title