Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 12 Antibody (SR1957), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32239850UL
Description
Cytokeratin 12 Monoclonal antibody specifically detects Cytokeratin 12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
Cytokeratin 12 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1957 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
keratin, type I cytoskeletal 12, CK12, K12, keratin 12 | |
A synthesized peptide derived from human Cytokeratin 12 (Uniprot #: Q99456) | |
50 μg | |
Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
3859 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction