Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytokeratin 12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565979
Description
Immunogen sequence: QGDGLEESLFV TDSKSQAQST DSSKDPTKTR K Highest antigen sequence identity to the following orthologs - mouse 63%, rat 72%.
KRT12 encodes the type I intermediate filament chain keratin 12, expressed in corneal epithelia. Mutations in this gene lead to Meesmann corneal dystrophy.
Specifications
Cytokeratin 12 | |
Polyclonal | |
Unconjugated | |
KRT12 | |
AI835216; CK-12; cytokeratin-12; K12; Ka12; keratin 12; keratin 12 (Meesmann corneal dystrophy); keratin 12, type I; keratin complex 1, acidic, gene 12; keratin K12; keratin, type I cytoskeletal 12; Keratin, type I cytoskeletal 12 (Cytokeratin 12); keratin-12; Krt1-12; Krt12; Krt-12; krt12 {ECO:0000250; Type I keratin Ka12; UniProtKB:Q99456} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3859 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99456 | |
KRT12 | |
Recombinant Human Cytokeratin 12. Recombinant protein control fragment (Product #RP-106668). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction