Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 12 Recombinant Rabbit Monoclonal Antibody (JE55-35)
GREENER_CHOICE

Catalog No. PIMA542505
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542505 Supplier Invitrogen™ Supplier No. MA542505
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: Mouse eyeball tissue lysates, rat eyeball tissue. Subcellular Location: Intermediate filament, cytosol, extracellular exosome.

KRT12 encodes the type I intermediate filament chain keratin 12, expressed in corneal epithelia. Mutations in this gene lead to Meesmann corneal dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE55-35
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene KRT12
Gene Accession No. Q64291, Q6IFW5, Q99456
Gene Alias AI835216; CK-12; cytokeratin-12; K12; Ka12; keratin 12; keratin 12 (Meesmann corneal dystrophy); keratin 12, type I; keratin complex 1, acidic, gene 12; keratin K12; keratin, type I cytoskeletal 12; Keratin, type I cytoskeletal 12 (Cytokeratin 12); keratin-12; Krt1-12; Krt12; Krt-12; krt12 {ECO:0000250; Type I keratin Ka12; UniProtKB:Q99456}
Gene Symbols KRT12
Host Species Rabbit
Immunogen Recombinant protein within C-terminal Human Keratin 12/K12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268482, 360625, 3859
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.