Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 13 Antibody (KS-1A3), Novus Biologicals™
SDP

Catalog No. NBP122777 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122777 1 mL
NB018465 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP122777 Supplier Novus Biologicals Supplier No. NBP122777
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Cytokeratin 13 Monoclonal specifically detects Cytokeratin 13 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 13
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KS-1A3
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry 1:25-1:100, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:25-1:100
Formulation Ascites with 0.05% Sodium Azide
Gene Alias CK13, CK-13, cytokeratin 13, Cytokeratin-13, K13cytokeratin-13, keratin 13, keratin, type I cytoskeletal 13, keratin-13, MGC161462, MGC3781
Gene Symbols KRT13
Host Species Mouse
Immunogen Cultured A431 cells from a human epidermoid carcinoma of the vulva.
Purification Method Ascites
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 3860
Test Specificity This is specific to a 54 kD protein. Cytokeratin 13 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 4. Cytokeratin 13 is expressed in major components of squamous, non-keratinized epithelium, transitional epithelium, and myoepithelium. This antibody is useful for identifying carcinomas of trachea, sweat glands, bladder, extocervix, tongue, esophagus, anal canal, and the basal layer of keratinized epidermis.
Target Species Human
Content And Storage Store at 4°C.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.