Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 13 Monoclonal Antibody (3G4)
GREENER_CHOICE

Catalog No. PIMA517109
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517109 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517109 Supplier Invitrogen™ Supplier No. MA517109
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17109 targets KRT13 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17109 immunogen is purified recombinant fragment of human KRT13 (amino acids: 143-295) expressed in E. Coli. MA5-17109 detects KRT13 which has a predicted molecular weight of approximately 52kDa.

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ~40 kDa and ~68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 13
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene KRT13
Gene Accession No. P13646
Gene Alias 47 kDa cytokeratin; CK13; CK-13; cytokeratin 13; cytokeratin-13; K13; K-13; Ka13; keratin 13; keratin 13, type I; keratin complex 1, acidic, gene 13; keratin, type I cytoskeletal 13; Keratin-13; Krt-1.13; Krt1-13; KRT13; krt13 {ECO:0000250; MGC161462; MGC3781; Type I keratin Ka13; UniProtKB:P13646}; WSN2
Gene Symbols KRT13
Host Species Mouse
Immunogen Purified recombinant fragment of human KRT13 (amino acids: 143-295) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3860
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.