Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 14 Antibody (KRT14/2375), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308531AF594
Description
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human, Mouse, Rat, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
Cytokeratin 14 | |
Monoclonal | |
Alexa Fluor 594 | |
CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ | |
Recombinant human Cytokeratin 14 fragment (around aa351-472) (exact sequence is proprietary) (Uniprot: P02533) | |
100 μg | |
Cell Biology, Cellular Markers | |
3861 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
KRT14/2375 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction