Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 14 Antibody (KRT14/4132), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB381967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381967 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381967 Supplier Novus Biologicals Supplier No. NBP308289AF488
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Cytokeratin 14
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone KRT14/4132
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ
Host Species Mouse
Immunogen Recombinant fragment (around aa351-472) of human Cytokeratin 14 protein (exact sequence is proprietary) (Uniprot: P02533 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3861
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.