Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 14 Antibody (KRT14/4132), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308289AF750
Description
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
Cytokeratin 14 | |
Monoclonal | |
Alexa Fluor 750 | |
CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ | |
Recombinant fragment (around aa351-472) of human Cytokeratin 14 protein (exact sequence is proprietary) (Uniprot: P02533 ) | |
100 μg | |
Cell Biology, Cellular Markers | |
3861 | |
Store at 4°C in the dark. | |
IgG2c κ |
Immunohistochemistry (Paraffin), Protein Array | |
KRT14/4132 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction