Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 14 Antibody (rKRT14/7368), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320619100UG
Description
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Cytokeratin 14 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ | |
Recombinant human Cytokeratin 14 fragment (around aa350-472) (exact sequence is proprietary) | |
100 μg | |
Cell Biology, Cellular Markers | |
3861 | |
Store at 4&drg;C. Do not freeze. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rKRT14/7368 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction