Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 14 Mouse anti-Human, Mouse, Rat, Porcine, Clone: KRT14/2375, Novus Biologicals™
SDP

Catalog No. p-7235114 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122674 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122674 Supplier Novus Biologicals Supplier No. NBP30760020UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 14 Monoclonal specifically detects Cytokeratin 14 in Human, Mouse, Rat, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 14
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT14/2375
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ
Host Species Mouse
Immunogen Recombinant human Cytokeratin 14 fragment (around aa351-472) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3861
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.