Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 14 Mouse, Clone: LL002, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$268.00 - $524.00
Specifications
Antigen | Cytokeratin 14 |
---|---|
Clone | LL002 |
Dilution | Western Blot 0.5-1.0ug/ml, Simple Western 10 ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 1-2ug/ml, Immunohistochemistry-Frozen 1-2ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP234270F
![]() |
Novus Biologicals
NBP2342700.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP234270A
![]() |
Novus Biologicals
NBP2342700.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP234270B
![]() |
Novus Biologicals
NBP2342700.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Cytokeratin 14 Monoclonal specifically detects Cytokeratin 14 in Human, Mouse, Rat samples. It is validated for Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Cytokeratin 14 | |
Western Blot 0.5-1.0ug/ml, Simple Western 10 ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 1-2ug/ml, Immunohistochemistry-Frozen 1-2ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
P02533 | |
3861 | |
A synthetic peptide of 15 amino acid from the C-terminus of human keratin 14 | |
Primary | |
Store at 4C. | |
50 kDa |
LL002 | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cytoskeleton Markers | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ | |
KRT14 | |
IgG3 | |
Protein A purified | |
Cytokeratin 14 (CK14) belongs to the type I (or A or acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5 (type II or B or basic). CK14 is found in basal cells of squamous epithelia, some glandular epithelia, myoepithelium, and mesothelial cells. Anti-CK14 is useful in differentiating squamous cell carcinomas from poorly differentiated epithelial tumors. Anti-CK14 is one of the specific basal markers for distinguishing between basal and non-basal subtypes of breast carcinomas. Anti-CK14 is also a good marker for differentiation of intraductal from invasive salivary duct carcinoma by the positive staining of basal cells surrounding the in-situ neoplasm as well as for differentiation of benign prostate from prostate carcinoma. Furthermore, this antibody has been useful in separating oncocytic tumors of the kidney from its renal mimics, and in identifying metaplastic carcinomas of the breast. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title