Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 15 Antibody (KRT15/2103R), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308609PE
Description
Cytokeratin 15 Monoclonal antibody specifically detects Cytokeratin 15 in Human, Rat, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
Cytokeratin 15 | |
Monoclonal | |
PE | |
CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15 | |
Recombinant human full-length Cytokeratin 15 protein (Uniprot: P19012) | |
100 μg | |
Cell Biology, Cellular Markers | |
3866 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
KRT15/2103R | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Rat, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction