Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 15 Antibody (LHK15), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$269.00 - $524.00

Specifications

Antigen Cytokeratin 15
Clone LHK15
Concentration 0.2 mg/mL
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1 ug/ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml, SDS-Page
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25330820
SDP
View Documents
Novus Biologicals
NBP25330820UG
20 μg
Each for $269.00
Only null left
Add to Cart
 
NB253308100
SDP
View Documents
Novus Biologicals
NBP253308100UG
100 μg
Each for $524.00
Only null left
Add to Cart
 
NB253308200
SDP
View Documents
Novus Biologicals
NBP253308200UG
200 μg N/A N/A N/A
Description

Description

Cytokeratin 15 Monoclonal specifically detects Cytokeratin 15 in Human, Rat, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Cytokeratin 15
0.2 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Mouse
Cytoskeleton Markers
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
3866
A 17 amino acids synthetic peptide from the C- terminal of Human Cytokeratin 15 (exact sequence is proprietary)
Primary
Store at 4C.
LHK15
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1 ug/ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml, SDS-Page
Monoclonal
Purified
RUO
Human, Rat, Bovine
CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15
KRT15
IgG2a κ
Protein A or G purified
52 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.