Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytokeratin 15 Monoclonal Antibody (LHK15)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA190929
Description
Recommended positive controls: Skin.
Cytokeratin 15 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21. 2.
Specifications
Cytokeratin 15 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P19012, Q61414, Q6IFV3 | |
KRT15 | |
A 17-mer synthetic peptide from the C-terminus of Keratin 15. | |
500 μL | |
Primary | |
Human, Mouse, Rat, Bovine | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry | |
LHK15 | |
Unconjugated | |
KRT15 | |
AI528832; C K1CO; CK15; CK-15; cytokeratin 15; cytokeratin-15; K15; K1CO; Ka15; keratin 15; keratin 15, type I; keratin complex 1, acidic, gene 15; keratin, type I cytoskeletal 15; Keratin-15; keratin-15, basic; keratin-15, beta; KRT1; Krt1-15; KRT15; krt15 {ECO:0000250; KRTB; type I cytoskeletal 15; type I keratin KA15; UniProtKB:P19012} | |
Mouse | |
Protein A | |
RUO | |
16665, 287700, 3866, 507464 | |
4°C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction