Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 15 Mouse anti-Human, Mouse, Clone: 6B4F8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Cytokeratin 15 |
---|---|
Clone | 6B4F8 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6345
![]() ![]() |
Proteintech Group Inc
602471IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Cytokeratin 15 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21.2.Specifications
Cytokeratin 15 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CK 15, CK15, Cytokeratin 15, K15, K1CO, keratin 15, KRT15, KRTB | |
KRT15 | |
IgG1 | |
Protein G | |
Antibody |
6B4F8 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P19012, Q61414 | |
16665, 3866 | |
Cytokeratin 15 Fusion Protein Ag0185 | |
Primary | |
-20°C | |
KRT15 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title