Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582887 Supplier Invitrogen™ Supplier No. PA582887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTTFLQTSSS TFGGGSTRGG SLLAGGGGFG GGSLSGGGGS RSISASSARF VSSGSGGGYG Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 83%.

Cytokeratin 15 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21. 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT15
Gene Accession No. P19012
Gene Alias AI528832; C K1CO; CK15; CK-15; cytokeratin 15; cytokeratin-15; K15; K1CO; Ka15; keratin 15; keratin 15, type I; keratin complex 1, acidic, gene 15; keratin, type I cytoskeletal 15; Keratin-15; keratin-15, basic; keratin-15, beta; KRT1; Krt1-15; KRT15; krt15 {ECO:0000250; KRTB; type I cytoskeletal 15; type I keratin KA15; UniProtKB:P19012}
Gene Symbols KRT15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRT15. Recombinant protein control fragment (Product #RP-93405).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3866
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.