Learn More
Invitrogen™ Cytokeratin 15 Polyclonal Antibody

Description
Antibody detects endogenous levels of total Cytokeratin 15.
Specifications
Specifications
| Antigen | Cytokeratin 15 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | KRT15 |
| Gene Accession No. | P19012, Q61414, Q6IFV3 |
| Gene Alias | AI528832; C K1CO; CK15; CK-15; cytokeratin 15; cytokeratin-15; K15; K1CO; Ka15; keratin 15; keratin 15, type I; keratin complex 1, acidic, gene 15; keratin, type I cytoskeletal 15; Keratin-15; keratin-15, basic; keratin-15, beta; KRT1; Krt1-15; KRT15; krt15 {ECO:0000250; KRTB; type I cytoskeletal 15; type I keratin KA15; UniProtKB:P19012} |
| Gene Symbols | KRT15 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.