Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 16 Antibody (OTI2H8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271085H
Description
Cytokeratin 16 Monoclonal specifically detects Cytokeratin 16 in Human, Rat samples. It is validated for Immunohistochemistry.Specifications
Cytokeratin 16 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
KRT16 | |
Full length recombinant protein of human Cytokeratin 16 (NP_005548) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG2a |
Immunohistochemistry | |
OTI2H8 | |
Immunohistochemistry | |
CK16, CK-16, cytokeratin 16, cytokeratin-16, FNEPPK, focal non-epidermolytic palmoplantar keratoderma, K16, K1CP, keratin 16, keratin, type I cytoskeletal 16, keratin-16, KRT16A, NEPPK | |
Mouse | |
Affinity Purified | |
Cytoskeleton Markers | |
3868.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction