Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytokeratin 16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597908
Description
Cytokeratin 16 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
KRT16 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region of chromosome 17q12-q21. This keratin has been coexpressed with keratin 14 in a number of epithelial tissues, including esophagus, tongue, and hair follicles. Mutations in this gene are associated with type 1 pachyonychia congenita, non-epidermolytic palmoplantar keratoderma and unilateral palmoplantar verrucous nevus.
Specifications
Cytokeratin 16 | |
Polyclonal | |
Unconjugated | |
KRT16 | |
AI324768; CK16; CK-16; cytokeratin 16; cytokeratin-16; FNEPPK; focal non-epidermolytic palmoplantar keratoderma; K16; K1CP; Ka16; keratin 16; keratin 16, type I; keratin complex 1, acidic, gene 16; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin, type I cytoskeletal 16; Keratin-16; Krt1-16; Krt16; KRT16A; NEPPK; PC1; type I keratin KA16 | |
Rabbit | |
Protein G | |
RUO | |
3868 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2.89 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P08779 | |
KRT16 | |
Recombinant Human Keratin, type I cytoskeletal 16 protein (265-333AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction