Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 17 Monoclonal Antibody (83-13), Huabio

Catalog No. 502544506
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4506 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4506 Supplier HUABIO RESEARCH INC Supplier No. M040713
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Cytokeratin 17
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 83-13
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene KRT17
Gene Accession No. Q04695, Q6IFU8, Q9QWL7
Gene Alias 39.1; CK17; CK-17; cytokeratin-17; hard keratin; K17; Ka17; keratin 17; keratin 17, type I; keratin complex 1, acidic, gene 17; keratin, type I cytoskeletal 17; Keratin-17; Krt1-17; Krt17; krt17 {ECO:0000250; PC; PC2; PCHC1; Type I keratin Ka17; UniProtKB:Q04695}
Host Species Mouse
Immunogen Synthetic peptide within Human Cytokeratin 17 aa 383-432/432.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16667, 287702, 3872
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.