Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 18 Antibody (C-04), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500306PE
Description
Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Mouse, Mammal samples. It is validated for Flow Cytometry.Specifications
Cytokeratin 18 | |
Monoclonal | |
PE | |
Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
The antibody C-04 reacts with Cytokeratin 18 (45 kDa), a member of intermediate filaments subfamily represented in epithelial tissues. | |
Store at 4C in the dark. |
Flow Cytometry | |
C-04 | |
Flow Cytometry | |
KRT18 | |
Cytoskeleton preparation of epidermal carcinoma cell line A431. | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Phospho Specific | |
3875 | |
Human, Mouse, Mammalia | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction