Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 18 Antibody (Cocktail), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247978MFV610
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human samples. It is validated for Flow CytometrySpecifications
Cytokeratin 18 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
Cocktail | |
Flow Cytometry | |
Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
Recombinant full-length human Cytokeratin 18 protein (Uniprot: P05783) | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
3875 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction