Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 18 Antibody (KRT18/1190) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$646.00

Specifications

Antigen Cytokeratin 18
Clone KRT18/1190
Concentration 1.0 mg/mL
Dilution Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP247984A
SDP
View Documents
Novus Biologicals
NBP2479840.1MG
0.1 mg
Each for $646.00
Only null left
Add to Cart
 
NBP247984B
SDP
View Documents
Novus Biologicals
NBP2479840.2MG
0.2 mg N/A N/A N/A
Description

Description

Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.
Specifications

Specifications

Cytokeratin 18
1.0 mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Unconjugated
Mouse
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers
PBS with No Preservative
3875
Recombinant human KRT18 protein
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
45 kDa
KRT18/1190
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human, Rat
Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18
KRT18
IgG1 κ
Protein A or G purified
This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It was reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.