Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 18 Antibody (KRT18/2808R), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308634IR
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Cytokeratin 18 | |
Monoclonal | |
DyLight 755 | |
Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
Recombinant human full-length Cytokeratin 18 protein (Uniprot: P05783) | |
100 μg | |
Cancer, Cell Biology, Cellular Markers | |
3875 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
KRT18/2808R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction