Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 18 Antibody (LC18N), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250462G
Description
Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Cytokeratin 18 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KRT18 | |
Synthetic pepide of N terminus of human keratin 18 (NH2- SFTTRSTFSC) | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers | |
3875 | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
LC18N | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Does not cross react with mouse. Cross reacts with keratin 15 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction