Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 18 Antibody (OTI1E1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271086R
Description
Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Cytokeratin 18 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KRT18 | |
Recombinant protein fragment corresponding to amino acids 69-372 of human Cytokeratin 18 (NP_000215) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1E1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
Mouse | |
Affinity Purified | |
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Phospho Specific | |
3875.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction