Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 18 Monoclonal Antibody (A4B4), Huabio

Catalog No. 502544523
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4523 Supplier HUABIO RESEARCH INC Supplier No. HA600020
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Cytokeratin 18
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A4B4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene KRT18
Gene Accession No. P05783
Gene Alias cb83; Cell proliferation-inducing gene 46 protein; CK 18; CK18; CK-18; CYK18; cytokeratin 18; cytokeratin endo B; cytokeratin type -1; DAPK-1; Cytokeratin18; cytokeratin-18; dapk1; dorsal aorta proneprin kinesin-1; DreK18; Endo B; I79_023185; K 18; K18; keratin 18; keratin 18, type I; keratin 18a, tandem duplicate 1; keratin complex 1, acidic, gene 18; keratin D; Keratin, type I cytoskeletal 18; Keratin18; keratin-18; Kerd; KRT 18; Krt1-18; KRT18; krt18 {ECO:0000250; krt18a.1; LOC100008885; PIG46; sb:cb83; type I cytoskeletal 18; UniProtKB:P05783}; wu:fa13f02; wu:fb36b04
Host Species Mouse
Immunogen Recombinant protein within human Cytokeratin 18 aa 1-150.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3875
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.