Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 19 Antibody (963410), Alexa Fluor 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB35062AF594
Description
Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cytokeratin 19 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
KRT19 | |
E.coli-derived recombinant human Cytokeratin 19, Gln311-Lys370, Accession No. P08727 | |
Primary | |
Human | |
IgG2b |
Western Blot, Immunohistochemistry | |
963410 | |
Western Blot, Immunohistochemistry | |
CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
Mouse | |
0.1 mL | |
3880 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction