Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 19 Antibody (KRT19/799) - IHC-Prediluted, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP248086
Description
Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 19 | |
Monoclonal | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
KRT19 | |
Recombinant human Cytokeratin 19 protein (Uniprot: P08727) | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a protein of 40kDa, identified as cytokeratin-19 (CK19), which is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer. | |
Store at 4C. | |
IgG2a λ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
KRT19/799 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
Mouse | |
40 kDa | |
7 mL | |
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuroscience, Stem Cell Markers | |
3880 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction