Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 19 Antibody (KRT19/800) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | Cytokeratin 19 |
---|---|
Clone | KRT19/800 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247946A
![]() |
Novus Biologicals
NBP2479460.1MG |
0.1 mg |
Each for $646.00
|
|
|||||
NBP247946B
![]() |
Novus Biologicals
NBP2479460.2MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Immunofluorescence, CyTOF-ready.Specifications
Cytokeratin 19 | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuroscience, Stem Cell Markers | |
PBS with No Preservative | |
3880 | |
Recombinant human KRT19 protein | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
40 kDa |
KRT19/800 | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
KRT19 | |
IgG1 κ | |
Protein A or G purified | |
Recognizes a protein of 40kDa, identified as cytokeratin-19 (CK19), which is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title