Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 19 Antibody (OTI3F8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271092F
Description
Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Cytokeratin 19 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
KRT19 | |
Recombinant fragment expressed in E.coli corresponding to amino acids 240-390 of human Cytokeratin 19. | |
0.1 mL | |
Primary | |
Human, Rat, Canine, Primate, Monkey | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3F8 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
Mouse | |
Affinity Purified | |
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuroscience, Stem Cell Markers | |
3880.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction