Learn More
Cytokeratin 19 Monoclonal Antibody (BA17), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$345.00
Specifications
Antigen | Cytokeratin 19 |
---|---|
Clone | BA17 |
Concentration | 0.5 mg/mL |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
53-989-882
![]() ![]() |
Invitrogen™
53989882 |
100 μg |
$460.00
$345.00 / Each of 1 Save $115.00 25% Off |
![]() |
|||||
Description
Description: This BA17 monoclonal antibody reacts with human cytokeratin 19 (CK19), a 44-kDa type I (acidic) intermediate filament protein that lacks the non-alpha-helical tail domain present in other keratins. Cytokeratins form the intracellular cytoskeletal network that maintains the integrity and stability of cells and tissues. Cytokeratin 19 is expressed in a wide variety of simple and stratified epithelial tissue. Moreover, cytokeratin 19 expression can be induced by vitamin A, SV40 transformation, and cancer. A soluble form of cytokeratin 19 generated by caspase 3 cleavage has also been found to be secreted by cancer cells, thus possibly indicating tumor metastasis. Cytokeratin 19 often exists as a heterodimer with cytokeratin 7, a type II keratin. Applications Reported: This BA17 antibody has been reported for use in immunohistochemical staining and immunocytochemistry. Applications Tested: This BA17 antibody has been tested by immunocytochemistry on fixed and permeabilized cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.Specifications
Cytokeratin 19 | |
0.5 mg/mL | |
Alexa Fluor 488 | |
Mouse | |
Human | |
3880 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
KRT19 |
BA17 | |
Monoclonal | |
Liquid | |
RUO | |
P08727 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.