Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 19 Monoclonal Antibody (G019.1), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 34059100UL

This item is not returnable.
View return policy
Description
Cytokeratin 19 Monoclonal antibody specifically detects Cytokeratin 19 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Cytokeratin 19 | |
Monoclonal | |
0.5-1.0 mg/mL | |
tris with 1% BSA and 0.1% sodium azide; pH 7.3-7.7 | |
P08727 | |
Mouse | |
Protein A | |
RUO | |
3880 | |
2-8°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
G019.1 | |
Unconjugated | |
KRT19 | |
40-kDa keratin intermediate filament; AI663979; C K1CS; CK 19; CK19; CK-19; cytokeratin 19; cytokeratin-19; EndoC; K19; K1CS; Ka19; keratin 19; keratin 19, type I; keratin complex 1, acidic, gene 19; keratin, type I cytoskeletal 19; keratin, type I, 40-kd; keratin-19; Krt-1.19; Krt1-19; Krt19; LOC100344434; MGC15366; Type I keratin Ka19 | |
Recombinant human Cytokeratin 19. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction