Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cytokeratin 20 Ab-1, Mouse Monoclonal Antibody

Catalog No. MS377S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS377S0 100 μL Concentrated
MS377S1 500 μL Concentrated
MS377S 1 mL Concentrated
MS377R7 7 mL Prediluted
MS377RQ 12 mL Ready-to-Use
MS377R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS377S0 Supplier Epredia™ Supplier No. MS377S0
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cytokeratin 20 Ab-1, Mouse Monoclonal Antibody.

Keratin 20/cytokeratin20 (CK20) is a Type-I keratin which is primarily expressed in gastric and intestinal epithelium, urothelium, and Merkel-cells. CK20 is expressed in adenocarcinomas of the colon, stomach, pancreas and the bile system. CK20 is also present in mucinous ovarian tumors, transitional-cell and Merkel-cell carcinomas. Notably, the squamous cell carcinomas and adenocarcinomas of the breast, lung, and endometrium, non-mucinous tumors of the ovary, and small cell carcinomas lack in CK20.

Host Species: Mouse

Clone: Ks20.8

Isotype: IgG2a

Species Reactivity: Human and Mouse. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Semi-purified human cytokeratin preparation

Molecular Weight: 46 kDa

Positive Control: HT29 cells, Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen Cytokeratin 20 Ab-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone Ks20.8
Conjugate Unconjugated
Host Species Mouse
Immunogen Semi-purified human cytokeratin preparation
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.