Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 20 Antibody (10D8), Alexa Fluor™ 647, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242616AF647

Catalog No. NBP242616AH


Only null left
Add to Cart

Description

Description

Cytokeratin 20 Monoclonal specifically detects Cytokeratin 20 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Cytokeratin 20
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
KRT20
Recombinant human KRT20 (1-424aa) purified from E.coli.
100 μL
Cytoskeleton Markers
54474
Store at 4C in the dark.
IgG2a κ
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
10D8
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.