Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 20 Monoclonal Antibody (2C7B9)
GREENER_CHOICE

Catalog No. FERMA548757
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548757 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548757 Supplier Invitrogen™ Supplier No. MA548757
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 20 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 20
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2C7B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene KRT20
Gene Accession No. P35900
Gene Alias 9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT
Gene Symbols KRT20
Host Species Mouse
Immunogen Purified recombinant fragment of human KRT20 (AA: 1-150) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54474
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.