Learn More
Cytokeratin 20 Monoclonal Antibody (Ks20.8), Zeta
Mouse Monoclonal Antibody Cocktail
$456.96
Specifications
Antigen | Cytokeratin 20 |
---|---|
Clone | Ks20.8 |
Concentration | 100 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-221-5852
![]() |
ZETA CORPORATION
Z2065ML |
1 mL |
Each of 1 for $456.96
|
|
|||||
Description
A recommended positive control tissue for this product is Colon, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
Cytokeratin 20 Monoclonal antibody specifically detects Cytokeratin 20 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Cytokeratin 20 | |
100 μg/mL | |
Monoclonal | |
Liquid | |
RUO | |
tris with BSA, NP-40 and <0.1% sodium azide | |
9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT | |
Semi-purified human cytokeratin preparation | |
Primary | |
2-8°C | |
KRT20 |
Ks20.8 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
P35900 | |
54474 | |
IgG2a κ | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.