Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 20 Mouse anti-Human, Clone: KRT20/1991, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30728520UG
Description
Cytokeratin 20 Monoclonal specifically detects Cytokeratin 20 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
Cytokeratin 20 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT | |
Recombinant fragment of human Cytokeratin 20 protein (around aa 196-323) (exact sequence is proprietary) | |
20 μg | |
Cell Biology, Cellular Markers | |
54474 | |
Store at 4°C. | |
IgG1 κ |
Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
KRT20/1991 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction