Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 20 Antibody (OTI3G1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271962IR
Description
Cytokeratin 20 Monoclonal specifically detects Cytokeratin 20 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cytokeratin 20 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KRT20 | |
Full length recombinant protein of human Cytokeratin 20 (NP_061883) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3G1 | |
Western Blot, Immunohistochemistry | |
CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT | |
Mouse | |
Affinity Purified | |
Cytoskeleton Markers | |
54474.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction