Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582891 Supplier Invitrogen™ Supplier No. PA582891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASYLEKVRTL EQSNSKLEVQ IKQWYETNAP RAGRDYSAYY RQIEELRSQI KDAQLQNARC VLQIDNAKLA A Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 77%.

The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 20
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT20
Gene Accession No. P35900
Gene Alias 9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT
Gene Symbols KRT20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRT20. Recombinant protein control fragment (Product #RP-93444).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54474
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.