Learn More
Cytokeratin 20 Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 50013674

Description
Positive control: HT29 cells. Brain Cellular location: Cytoplasm.
Cytokeratin 20 Polyclonal antibody specifically detects Cytokeratin 20 in Human samples. It is validated for Western BlotSpecifications
Cytokeratin 20 | |
Polyclonal | |
Unconjugated | |
KRT20 | |
9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT | |
Recombinant protein encoding aa 230-425 of human keratin 20 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P35900 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54474 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.