Learn More
Invitrogen™ Cytokeratin 20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587390
Description
Immunogen sequence: KYEVMAQKNL QEAKEQFERQ TAVLQQQVTV NTEELKGTEV QLTELRRTSQ SLEIELQSHL SMKESLEHTL EETKARYSSQ LANLQSLLSS LEAQLMQIRS NMERQNNEYH ILLDIKTRLE QEIATYRRLL EGEDVKTTEY QLSTLEERDI KKTRKIKTVV QEVVDGKVVS SEVKEVEENI; Positive Samples: HeLa, Mouse intestine; Cellular Location: Cytoplasm.
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
Specifications
Cytokeratin 20 | |
Polyclonal | |
Unconjugated | |
KRT20 | |
9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54474, 66809 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35900, Q9D312 | |
KRT20 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 245-424 of human Cytokeratin 20 (Cytokeratin 20 (KRT20)) (NP_0618831). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.