Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 20 Rabbit anti-Human, Clone: KRT20/3129R, Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50
Specifications
| Antigen | Cytokeratin 20 |
|---|---|
| Clone | KRT20/3129R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Cytokeratin 20 Monoclonal specifically detects Cytokeratin 20 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 20 | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 54474 | |
| IgG | |
| Protein A or G purified |
| KRT20/3129R | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT | |
| Recombinant fragment of human Cytokeratin 20 protein (around aa 196-323) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title